2-(Methylsulphanyl)phenyl isocyanate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Isocyanatophenyl methyl sulphide, 1-Isocyanato-2-(methylthio)benzene, 2-Isocyanatothioanisole

OR0658-1G
APOSOR0658-1GEA 125 EUR
APOSOR0658-1G
2-(Methylsulphanyl)phenyl isocyanate 95+%
2-(Methylsulphanyl)phenyl isocyanate

May form small amount of sediment

Formel: C₈H₇NOS
Molecular Weight: 165,212 g/mol
Siedepunkt: 126 °C (5 mm Hg)
Dichte: 1,21 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037076
CAS-Nummer: 52260-30-7
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR