4-Morpholino-3-nitrobenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR0662-250MG
APOSOR0662-250MGEA
21.9
EUR
APOSOR0662-250MG
4-Morpholino-3-nitrobenzaldehyd
4-Morpholino-3-nitrobenzaldehyd
Formel:
C₁₁H₁₂N₂O₄ Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
300541-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...