Imidazo[2,1-b]thiazole-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR0694-250MG
APOSOR0694-250MGEA 40.6 EUR
APOSOR0694-250MG
Imidazo[2,1-b]thiazole-6-carboxylic acid
Imidazo[2,1-b]thiazole-6-carboxylic acid
Formel: C₆H₄N₂O₂S
Molecular Weight: 168,1732 g/mol
Schmelzpunkt: 233…234 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03419463
CAS-Nummer: 53572-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR