D-α-Phenylalanin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
D-Phenylalanin, (R)-(+)-2-Amino-3-phenylpropansäure, 3-Phenyl-D-alanin, H-D-Phe-OH, 3-Phenyl-D(+)-alanin, (R)-(+)-2-Amino-3-phenylpropionsäure, (R)-(+)-2-Amino-3-phenylpropionic acid
OR0702-5G
APOSOR0702-5GEA
58
EUR
APOSOR0702-5G
D-α-Phenylalanin 97%
D-α-Phenylalanin
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Schmelzpunkt: 273…276 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004270 CAS-Nummer: 673-06-3 EINECS: 211-603-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...