2-(1H-Imidazol-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Carboxyphenyl)-1H-imidazole, 2-(1-Imidazolyl)benzoic Acid

OR0715-250MG
APOSOR0715-250MGEA 43.5 EUR
APOSOR0715-250MG
2-(1H-Imidazol-1-yl)benzoic acid
2-(1H-Imidazol-1-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,183 g/mol
Schmelzpunkt: 198…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06254507
CAS-Nummer: 159589-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR