3-Morpholin-4-ylbenzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR0716-250MG
APOSOR0716-250MGEA
21.9
EUR
APOSOR0716-250MG
3-Morpholin-4-ylbenzoic acid 97%
3-Morpholin-4-ylbenzoic acid
Formel:
C₁₁H₁₃NO₃ Schmelzpunkt: 150…165 °C |
MDL:
MFCD06659078 CAS-Nummer: 215309-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...