D(+)-Prolin

Lieferant: Apollo Scientific
Achtung

OR0718-100G OR0718-25G
APOSOR0718-100GEA 0 EUR
APOSOR0718-100G APOSOR0718-25G
D(+)-Prolin
D(+)-Prolin

Chiral intermediate for pharmaceuticals and agrochemicals.

Formel: C₅H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR