(S)-(-)-Indolin-2-carbonsäure

Lieferant: Apollo Scientific

Synonyme: (S)-Indoline-2-carboxylic acid

OR0737-100G
APOSOR0737-100GEA 116 EUR
APOSOR0737-100G
(S)-(-)-Indolin-2-carbonsäure
(S)-(-)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070578
CAS-Nummer: 79815-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR