(p-Toluolsulfonyl)methylisocyanid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide

OR0752-25G
APOSOR0752-25GEA 21.9 EUR
APOSOR0752-25G
(p-Toluolsulfonyl)methylisocyanid
(p-Toluolsulfonyl)methylisocyanid
Formel: C₉H₉NO₂S
Molecular Weight: 195.24 g/mol
Schmelzpunkt: 109…114 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00000005
CAS-Nummer: 36635-61-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR