N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
Lieferant: Apollo Scientific
|
Achtung
|
OR0754-10MG
OR0754-50MG
APOSOR0754-10MGEA
115
EUR
APOSOR0754-10MG
APOSOR0754-50MG
N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide
Formel:
C₁₅H₁₅N₅O Molecular Weight: 281.312 g/mol Schmelzpunkt: 200…201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01567008 CAS-Nummer: 108825-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...