2,4,6-Tri-tert-butylanilin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,6-Tris(tert-butyl)aniline

OR0755-5G
APOSOR0755-5GEA 55.1 EUR
APOSOR0755-5G
2,4,6-Tri-tert-butylanilin 98%
2,4,6-Tri-tert-butylanilin
Formel: C₁₈H₃₁N
Molecular Weight: 261,45 g/mol
Schmelzpunkt: 144…149 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011645
CAS-Nummer: 961-38-6
EINECS: 213-507-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR