4-(Piperidin-1-yl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR0759-100MG
APOSOR0759-100MGEA
21.9
EUR
APOSOR0759-100MG
4-(Piperidin-1-yl)benzoic acid
4-(Piperidin-1-yl)benzoic acid
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205,2530 g/mol Schmelzpunkt: 214…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00587602 CAS-Nummer: 22090-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...