DL-Tryptophan methyl ester hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride, H-DL-Trp-OMe·HCl

OR0765-25G
APOSOR0765-25GEA 43.5 EUR
APOSOR0765-25G
DL-Tryptophan methyl ester hydrochloride 97%
DL-Tryptophan methyl ester hydrochloride
Formel: C₁₂H₁₅ClN₂O₂
Molecular Weight: 254.714 g/mol
Schmelzpunkt: 221…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066133
CAS-Nummer: 5619-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR