3-Piperidin-1-ylbenzylamine 95%

Lieferant: Apollo Scientific
Gefahr

OR0766-100MG
APOSOR0766-100MGEA 21.9 EUR
APOSOR0766-100MG
3-Piperidin-1-ylbenzylamine 95%
3-Piperidin-1-ylbenzylamine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659082
CAS-Nummer: 175696-71-6
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR