4-Phenyl-1-butanol 97%
Lieferant: Apollo Scientific
Synonyme:
4-Phenylbutan-1-ol
OR0769-100G
APOSOR0769-100GEA
65.4
EUR
APOSOR0769-100G
4-Phenyl-1-butanol 97%
4-Phenyl-1-butanol
Formel:
(C₆H₅)(CH₂)₃CH₂OH Molecular Weight: 150.22 g/mol Siedepunkt: 138…140 °C/14mmHg Dichte: 0.975 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002971 CAS-Nummer: 3360-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...