4-(Methylthio)benzaldehyd

Lieferant: Apollo Scientific

Synonyme: 4-(Methylmercapto)benzaldehyd

OR0776-100G
APOSOR0776-100GEA 56.7 EUR
APOSOR0776-100G
4-(Methylthio)benzaldehyd
4-(Methylthio)benzaldehyd
Formel: C₈H₈OS
Molecular Weight: 152,22 g/mol
Siedepunkt: 267 °C (1013 hPa)
Dichte: 1,14 g/cm³ (20 °C)
MDL: MFCD00006948
CAS-Nummer: 3446-89-7
EINECS: 222-365-7
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR