3,5-Dibromo-1-benzofuran
Lieferant: Apollo Scientific
|
Achtung
|
OR0797-1G
APOSOR0797-1GEA
183
EUR
APOSOR0797-1G
3,5-Dibromo-1-benzofuran
3,5-Dibromo-1-benzofuran
Formel:
C₈H₄Br₂O Schmelzpunkt: 76…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659085 CAS-Nummer: 99660-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...