1-(2-Aminoethyl)-4-methylpiperazin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR0800-5G
OR0800-10G
APOSOR0800-5GEA
55.1
EUR
APOSOR0800-5G
APOSOR0800-10G
1-(2-Aminoethyl)-4-methylpiperazin 95%
1-(2-Aminoethyl)-4-methylpiperazin
Formel:
C₇H₁₇N₃ Siedepunkt: 112 °C/30mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03701701 CAS-Nummer: 934-98-5 EINECS: 213-296-3 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...