3-Amino-1-methylpiperidine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methylpiperidin-3-amine dihydrochloride

OR0801-1G
APOSOR0801-1GEA 21.9 EUR
APOSOR0801-1G
3-Amino-1-methylpiperidine dihydrochloride
3-Amino-1-methylpiperidine dihydrochloride
Formel: C₆H₁₆Cl₂N₂
Molecular Weight: 187,11 g/mol
Schmelzpunkt: 236…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06796280
CAS-Nummer: 894808-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR