1-Boc-4-piperidinessigsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR0804-10G
OR0804-1G
OR0804-5G
APOSOR0804-10GEA
0
EUR
APOSOR0804-10G
APOSOR0804-1G
APOSOR0804-5G
1-Boc-4-piperidinessigsäure
1-Boc-4-piperidinessigsäure
Formel:
C₁₂H₂₁NO₄ Schmelzpunkt: 99…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800239 CAS-Nummer: 157688-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...