1-(Phenylsulphonyl)piperazine 98+%

Lieferant: Apollo Scientific
Achtung

OR0822-5G
APOSOR0822-5GEA 192 EUR
APOSOR0822-5G
1-(Phenylsulphonyl)piperazine 98+%
1-(Phenylsulphonyl)piperazine
Formel: C₁₀H₁₄N₂O₂S
Molecular Weight: 226.3 g/mol
Schmelzpunkt: 106…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02182804
CAS-Nummer: 14172-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR