3-Iodbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR0825-100G
OR0825-25G
OR0825-5G
APOSOR0825-100GEA
144
EUR
APOSOR0825-100G
APOSOR0825-25G
APOSOR0825-5G
3-Iodbenzonitril 98%
3-Iodbenzonitril
Formel:
C₇H₄IN Molecular Weight: 229.02 g/mol Schmelzpunkt: 39…44 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00079762 CAS-Nummer: 69113-59-3 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...