1,7-Naphthyridin-8-amine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Amino-1,7-naphthyridine

OR0830-250MG
APOSOR0830-250MGEA 36.4 EUR
APOSOR0830-250MG
1,7-Naphthyridin-8-amine 95%
1,7-Naphthyridin-8-amine
Formel: C₈H₇N₃
Molecular Weight: 145,17 g/mol
Schmelzpunkt: 167…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319219
CAS-Nummer: 17965-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR