tert-Butyl-4-formylbenzylcarbamate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Aminomethyl)benzaldehyde, N-BOC protected, tert-Butyl 4-formylbenzylcarbamate

OR0835-250MG
APOSOR0835-250MGEA 21.9 EUR
APOSOR0835-250MG
tert-Butyl-4-formylbenzylcarbamate 97%
tert-Butyl-4-formylbenzylcarbamate
Formel: C₁₃H₁₇NO₃
Schmelzpunkt: 82,9…83.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659090
CAS-Nummer: 156866-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR