Piperazine-1-carboxylic acid diphenylamide

Lieferant: Apollo Scientific
Achtung

OR0836-1G
APOSOR0836-1GEA 174 EUR
APOSOR0836-1G
Piperazine-1-carboxylic acid diphenylamide
Piperazine-1-carboxylic acid diphenylamide
Formel: C₁₇H₁₉N₃O
Molecular Weight: 281,352 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973347
CAS-Nummer: 1804-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR