1-(Methylsulfonyl)piperazin

Lieferant: Apollo Scientific

OR0841-5G OR0841-25G OR0841-100G
APOSOR0841-5GEA 0 EUR
APOSOR0841-5G APOSOR0841-25G APOSOR0841-100G
1-(Methylsulfonyl)piperazin
1-(Methylsulfonyl)piperazin
Formel: C₅H₁₂N₂O₂S
MDL: MFCD02153225
CAS-Nummer: 55276-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR