[4-(Dimethylamino)phenyl]methanol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxymethyl-N,N-dimethylaniline

OR0847-1G OR0847-250MG OR0847-5G
APOSOR0847-1GEA 95.7 EUR
APOSOR0847-1G APOSOR0847-250MG APOSOR0847-5G
[4-(Dimethylamino)phenyl]methanol 97%
[4-(Dimethylamino)phenyl]methanol
Formel: C₉H₁₃NO
Molecular Weight: 151.21 g/mol
Siedepunkt: 125…126 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03840449
CAS-Nummer: 1703-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR