1-Adamantan-1-ylpiperazine
Lieferant: Apollo Scientific
|
Achtung
|
OR0860-1G
OR0860-5G
APOSOR0860-1GEA
0
EUR
APOSOR0860-1G
APOSOR0860-5G
1-Adamantan-1-ylpiperazine
1-Adamantan-1-ylpiperazine
Formel:
C₁₄H₂₄N₂ Schmelzpunkt: 87…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01748381 CAS-Nummer: 19984-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...