Phenyl(1-piperazinyl)methanon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Phenyl(piperazin-1-yl)methanone
OR0862-1G
APOSOR0862-1GEA
21.9
EUR
APOSOR0862-1G
Phenyl(1-piperazinyl)methanon
1-Benzoylpiperazine
Formel:
C₁₁H₁₄N₂O Molecular Weight: 190,24 g/mol Siedepunkt: 145 °C Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00810192 CAS-Nummer: 13754-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...