1-Boc-3-hydroxypiperidin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-butyl 3-hydroxypiperidine-1-carboxylate
OR0870-25G
OR0870-5G
OR0870-100G
APOSOR0870-25GEA
40.6
EUR
APOSOR0870-25G
APOSOR0870-5G
APOSOR0870-100G
1-Boc-3-hydroxypiperidin
1-Boc-3-hydroxypiperidin
Formel:
C₁₀H₁₉NO₃ Schmelzpunkt: 65…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093938 CAS-Nummer: 85275-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...