4-(Dipropylamino)piperidine 98%

Lieferant: Apollo Scientific
Gefahr

OR0877-500MG
APOSOR0877-500MGEA 0 EUR
APOSOR0877-500MG
4-(Dipropylamino)piperidine 98%
4-(Dipropylamino)piperidine
Formel: C₁₁H₂₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04117822
CAS-Nummer: 675136-96-6
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR