N,N-Dimethylpiperazine-1-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Dimethylcarbamoyl)piperazine

OR0897-1G
APOSOR0897-1GEA 43.5 EUR
APOSOR0897-1G
N,N-Dimethylpiperazine-1-carboxamide
N,N-Dimethylpiperazine-1-carboxamide
Formel: C₇H₁₅N₃O
Molecular Weight: 157,213 g/mol
Siedepunkt: 86 °C (0,3 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01050443
CAS-Nummer: 41340-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR