(R)-(-)-2-Amino-1-propanol

Lieferant: Apollo Scientific
Gefahr

Synonyme: D(-)-Alaninol, D(-)-2-Amino-1-propanol

OR0903-100G OR0903-25G
APOSOR0903-100GEA 174 EUR
APOSOR0903-100G APOSOR0903-25G
(R)-(-)-2-Amino-1-propanol
(R)-(-)-2-Amino-1-propanol

Chiral intermediate for pharmaceuticals and agrochemicals.

Formel: C₃H₉NO
Molecular Weight: 75.11 g/mol
Siedepunkt: 168…176 °C
Dichte: 0.965 g/cm³ (25 °C)
Flammpunkt: 83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064413
CAS-Nummer: 35320-23-1
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR