(3-Chlorpropyl)dimethylammoniumchlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR0910-500G
OR0910-100G
OR0910-1KG
APOSOR0910-500GEA
0
EUR
APOSOR0910-500G
APOSOR0910-100G
APOSOR0910-1KG
(3-Chlorpropyl)dimethylammoniumchlorid 97%
3-Chloropropyldimethylammonium chloride
Formel:
C₅H₁₃Cl₂N Schmelzpunkt: 138…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012521 CAS-Nummer: 5407-04-5 EINECS: 226-467-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...