5-Nitro-1H-indazol-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-nitro-1H-indazole

OR0925-1G
APOSOR0925-1GEA 0 EUR
APOSOR0925-1G
5-Nitro-1H-indazol-3-amine
5-Nitro-1H-indazol-3-amine
Formel: C₇H₆N₄O₂
Molecular Weight: 178,1500 g/mol
Schmelzpunkt: 254…256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179824
CAS-Nummer: 41339-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR