Ethyl-2-(3-piperidinyl)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl (piperidin-3-yl)acetate

OR0951-250MG OR0951-1G OR0951-5G
APOSOR0951-250MGEA 58 EUR
APOSOR0951-250MG APOSOR0951-1G APOSOR0951-5G
Ethyl-2-(3-piperidinyl)acetate
Ethyl-2-(3-piperidinyl)acetate
Formel: C₉H₁₇NO₂
Molecular Weight: 171.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412125
CAS-Nummer: 64995-88-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR