Ethyl-2-(3-piperidinyl)acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl (piperidin-3-yl)acetate
OR0951-250MG
OR0951-1G
OR0951-5G
APOSOR0951-250MGEA
58
EUR
APOSOR0951-250MG
APOSOR0951-1G
APOSOR0951-5G
Ethyl-2-(3-piperidinyl)acetate
Ethyl-2-(3-piperidinyl)acetate
Formel:
C₉H₁₇NO₂ Molecular Weight: 171.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412125 CAS-Nummer: 64995-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...