Ethyl-2-(piperidin-4-yl)acetate
Lieferant: Apollo Scientific
|
Achtung
|
OR0952-1G
APOSOR0952-1GEA
23
EUR
APOSOR0952-1G
Ethyl-2-(piperidin-4-yl)acetate
Formel:
C₉H₁₇NO₂ Siedepunkt: 72…73 °C/0,08mmHg Lagertemperatur: Gefrierschrank |
MDL:
MFCD02183575 CAS-Nummer: 59184-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...