2-Ethylphenylhydraziniumchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(2-Ethylphenyl)hydrazin Hydrochlorid, 2-Ethylphenylhydrazin Hydrochlorid
OR0985-100G
APOSOR0985-100GEA
156
EUR
APOSOR0985-100G
2-Ethylphenylhydraziniumchlorid 97%
2-Ethylphenylhydraziniumchlorid
Formel:
C₈H₁₃ClN₂ Molecular Weight: 172.66 g/mol Schmelzpunkt: 178 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071599 CAS-Nummer: 19398-06-2 EINECS: 421-460-2 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...