3-(Benzyldimethylammonio)propanesulphonate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
NDSB-256, 3-(Benzyldimethylammonio) propane-1-sulphonate, 3-[dimethylbenzylamino]propanesulphonic acid
OR1004-1G
APOSOR1004-1GEA
23.2
EUR
APOSOR1004-1G
3-(Benzyldimethylammonio)propanesulphonate 95+%
3-(Benzyldimethylammonio)propanesulphonate
Formel:
C₁₂H₁₉NO₃S Molecular Weight: 257,348 g/mol Schmelzpunkt: 245…248 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00225018 CAS-Nummer: 81239-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...