2-(4-Chlorophenyl)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-4-(piperidin-2-yl)benzene hydrochloride

OR1006-250MG
APOSOR1006-250MGEA 371 EUR
APOSOR1006-250MG
2-(4-Chlorophenyl)piperidine hydrochloride
2-(4-Chlorophenyl)piperidine hydrochloride
Formel: C₁₁H₁₄ClN·ClH
Molecular Weight: 232,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02178960
CAS-Nummer: 1172776-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR