δ-Valerolacton 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Pentansäure-δ-lacton, 5-Hydroxyvaleriansäurelacton, Tetrahydro-2H-pyran-2-on, Pentan-5-olid

OR10065-100G OR10065-1KG OR10065-500G
APOSOR10065-100GEA 21.9 EUR
APOSOR10065-100G APOSOR10065-1KG APOSOR10065-500G
δ-Valerolacton 99%
δ-Valerolacton

Material may contain up to 25% polymerised material which can be reverted to the product by distillation.

Formel: C₅H₈O₂
Molecular Weight: 100.12 g/mol
Siedepunkt: 58…60 °C/0.5mmHg
Schmelzpunkt: –13 °C
Dichte: 1.056 g/cm³ (25 °C)
Flammpunkt: 112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006645
CAS-Nummer: 542-28-9
EINECS: 208-807-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR