trans-4-Aminocyclohexanol Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR10070-25G OR10070-5G
APOSOR10070-25GEA 24.8 EUR
APOSOR10070-25G APOSOR10070-5G
trans-4-Aminocyclohexanol Hydrochlorid
trans-4-Aminocyclohexanol Hydrochlorid
Formel: C₆H₁₄ClNO
Molecular Weight: 151.64 g/mol
Schmelzpunkt: 225…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012566
CAS-Nummer: 50910-54-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR