4-Nitrophenyl-phenylsulfid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Nitrophenyl)(phenyl)sulphane
OR10071-1G
OR10071-25G
OR10071-5G
APOSOR10071-1GEA
21.9
EUR
APOSOR10071-1G
APOSOR10071-25G
APOSOR10071-5G
4-Nitrophenyl-phenylsulfid
4-Nitrophenyl-phenylsulfid
Formel:
C₁₂H₉NO₂S Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024700 CAS-Nummer: 952-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...