5-Benzyloxyindol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Benzyloxy-1H-indol

OR10082-1G
APOSOR10082-1GEA 21.9 EUR
APOSOR10082-1G
5-Benzyloxyindol
5-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
Schmelzpunkt: 96…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005676
CAS-Nummer: 1215-59-4
EINECS: 214-930-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR