5-Chloro-6-(3-chlorophenyl)-2-methylpyridazin-3(2H)-one 95+%

Lieferant: Apollo Scientific

OR1012-1G
APOSOR1012-1GEA 246 EUR
APOSOR1012-1G
5-Chloro-6-(3-chlorophenyl)-2-methylpyridazin-3(2H)-one 95+%
5-Chloro-6-(3-chlorophenyl)-2-methylpyridazin-3(2H)-one
Formel: C₁₁H₈Cl₂N₂O
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06800420
CAS-Nummer: 849021-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR