Benzo[b]thiophen-5-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[b]thiophene-5-carbaldehyde

OR101286-250MG
APOSOR101286-250MGEA 91.5 EUR
APOSOR101286-250MG
Benzo[b]thiophen-5-carbaldehyd
Benzo[b]thiophen-5-carbaldehyd
Formel: C₉H₆OS
Molecular Weight: 162.2083 g/mol
Schmelzpunkt: 56…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663673
CAS-Nummer: 10133-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR