5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-2,3-dihydro-1-benzofuran-7-sulphonyl chloride, 5-Bromo-2,3-dihydrobenzofuran-7-sulphonyl chloride, 5-Bromo-7-(chlorosulphonyl)-2,3-dihydrobenzo[b]furan
OR101632-1G
APOSOR101632-1GEA
328
EUR
APOSOR101632-1G
5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride
5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride
Formel:
C₈H₆BrClO₃S Molecular Weight: 297,553 g/mol Schmelzpunkt: 121…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04035726 CAS-Nummer: 690632-00-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...