5-Chlor-1,3,3-trimethyl-2-methylenindolin 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-2,3-dihydro-1,3,3-trimethyl-2-methylene-1H-indole

OR10170-25G
APOSOR10170-25GEA 151 EUR
APOSOR10170-25G
5-Chlor-1,3,3-trimethyl-2-methylenindolin 95%
5-Chlor-1,3,3-trimethyl-2-methylenindolin
Formel: C₁₂H₁₄ClN
Molecular Weight: 207.70 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005814
CAS-Nummer: 6872-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR