5-Indolcarbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Indole-5-carboxylic acid
OR10183-25G
OR10183-100G
APOSOR10183-25GEA
95.7
EUR
APOSOR10183-25G
APOSOR10183-100G
5-Indolcarbonsäure 98%
5-Indolcarbonsäure
Formel:
C₉H₇NO₂ Schmelzpunkt: 208…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005678 CAS-Nummer: 1670-81-1 EINECS: 216-799-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...